Order Entry
Austria
ContactUsLinkComponent
Di-tert-butyl pyrocarbonate for synthesis
Flammable
Toxic
Di-tert-butyl pyrocarbonate for synthesis
Supplier:  Merck
Certificates
undefined
Flammable
Toxic
Di-tert-butyl pyrocarbonate for synthesis
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    20 - 24 °C
  • Molar Mass:
    218.25 g/mol
  • Boiling Point:
    56 - 57 °C (0.7 hPa)
  • Flash Point:
    37 °C
  • Density:
    1.02 g/cm3 (20 °C)
  • Ignition Point:
    460 °C
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic Materials

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together