Order Entry
Austria
ContactUsLinkComponent
3-Octanone, Sigma-Aldrich®
3-Octanone, Sigma-Aldrich®
Catalog # SIAL136913-25G
Supplier:  Merck
CAS Number:  
3-Octanone, Sigma-Aldrich®
Catalog # SIAL136913-25G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :128.21 g/mol
Merck Index:13,03799
Boiling Pt:168 °C (1013 hPa)
Melting Pt:–18.5 °C
Density:0.82 g/cm³ (20 °C)
Flash Pt:43 °C
Storage Temperature:Refrigerator
EINECS:203-423-0
UN:1224
ADR:3,III
 :M-Clarity™ quality level = MQ100
CAS Number:106-68-3
Formula:C₈H₁₆O
MDL Number:MFCD00009515