Order Entry
Austria
ContactUsLinkComponent
Dichloroacetic acid (DCA)
Corrosive
Environment
Dichloroacetic acid (DCA)
Catalog # EHERC12320000
Supplier:  EHRENSTORFER
CAS Number:  
Corrosive
Environment
Dichloroacetic acid (DCA)
Catalog # EHERC12320000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Merck Index:13,03075
Boiling Pt:194 °C (1013 hPa)
Melting Pt:∼–4 °C
Density:1.563 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-207-0
UN:1764
ADR:8,II
 :128.94 g/mol
 :GHS05
 :GHS09
CAS Number:79-43-6
Formula:C₂H₂Cl₂O₂
MDL Number:MFCD00004223