Order Entry
Austria
ContactUsLinkComponent
Dichloromethane
Health
Irritant
Dichloromethane
Catalog # ROTHP089.1
Supplier:  Roth Carl
CAS Number:  
undefined
Health
Irritant
Dichloromethane
Catalog # ROTHP089.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

About this item

 :
 :84.93 g/mol
Merck Index:12,06140
Boiling Pt:39.8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1.322 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-838-9
UN:1593
ADR:6.1,III
 :GHS08
 :GHS07
CAS Number:75-09-2
Formula:CH₂Cl₂
MDL Number:MFCD00000881