Order Entry
Austria
Orders LinkContactUsLinkComponent
Hexanal, Sigma-Aldrich®

Hexanal, Sigma-Aldrich®

Flammable
Irritant
Supplier:   Merck
Hexanal, Sigma-Aldrich®
Supplier:  Merck
Certificates
Hexanal, Sigma-Aldrich®

3 Options Available Below

Product Details & Documents
Synonyms:  Hexylaldehyde, Caproaldehyde, Hexaldehyde, Aldehyde C6, Hexyl aldehyde
  • CAS Number:66-25-1
  • Formula:C₆H₁₂O
  • MDL Number:MFCD00007027
  • Molecular Weight:100.16 g/mol
  • Merck Index:13,01766
  • Boiling Pt:131 °C (1013 hPa)
  • Melting Pt:–56 °C
  • Density:0.81 g/cm³ (20 °C)
  • Flash Pt:25 °C
  • Storage Temperature:Refrigerator
  • UN:1207
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ100
  • EINECS:200-624-5
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 223,00 €
    223,00 €
    1 * 100 g
       223,00 €

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 1 460,00 €
    1 460,00 €
    1 * 1 kg
       1 460,00 €