Order Entry
Austria
ContactUsLinkComponent
Bronopol
Corrosive
Irritant
Environment
Bronopol
Catalog # EHERC10810000
Supplier:  EHRENSTORFER
CAS Number:  
Corrosive
Irritant
Environment
Bronopol
Catalog # EHERC10810000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :199.99 g/mol
Merck Index:13,01432
Boiling Pt:388 °C (1013 hPa)
Melting Pt:130…133 °C
Density:1.91 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-143-0
UN:3241
ADR:4.1,III
 :GHS05
 :GHS07
 :GHS09
CAS Number:52-51-7
Formula:HOCH₂BrC(NO₂)CH₂OH
MDL Number:MFCD00007390