1-Bromoheptane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:629-04-9
:
Formula:BrH₂C(CH₂)₅CH₃
MDL Number:MFCD00000273
:179.1 g/mol
Boiling Pt:179 °C (1013 hPa)
Melting Pt:–58 °C
Density:1.1602 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-068-8
ADR:3,III
:M-Clarity™ quality level = MQ200
UN:1993
Frequently Bought Together