Order Entry
Austria
ContactUsLinkComponent
pH/mV/°C meter, bench top, Eutech pH 2700
pH/mV/°C meter, bench top, Eutech pH 2700
Catalog # EUTEECPH270042GS
Supplier:  EUTECH
undefined
pH/mV/°C meter, bench top, Eutech pH 2700
Catalog # EUTEECPH270042GS
Supplier:  EUTECH

Specifications

  • Temperature range:
    0,0...+100 °C
  • Description:
    pH/mV/°C meter, bench top, Eutech pH 2700
  • Model:
    Eutech pH 2700
  • Temperature accuracy:
    ±0,3 °C
  • Temperature resolution:
    0,1 °C
  • Data logging capacity:
    500 data points
  • Display:
    Backlit
  • Outputs:
    RS232
  • Power supply:
    9 V DC adapter, 1,3 A (100/240 VAC, SMPS)
  • Temperature compensation:
    ATC / MTC
  • W×D×H:
    155×175×69 mm
  • mV accuracy:
    ±0,2
  • mV range:
    ±2000,0
  • mV resolution:
    0,1
  • pH accuracy:
    ±0,002 pH + 1 LSD
  • pH calibration:
    Up to 6 points (preset) or 5 points (custom)
  • pH range:
    −2,000...+20,000
  • pH resolution:
    0,1 / 0,01 / 0,001
  • Pk:
    1

Specifications

About this item

This bench meter comes with a large, one-glance-sees-all screen. Users can view pH or ORP readings, with temperature, electrode status, calibration points, date and time all at once.

  • Up to 6-point push button calibration with auto-buffer recognition
  • Quick, easy electrode diagnosis with pH slope and offset display
  • Non volatile memory holds up to 500 data points – time and date-stamped for GLP compliance
  • Cal-due alarm – no more out-dated calibrations
  • Auto-logging function for convenient continuous monitoring
  • Splashproof housing and keypad protects against incidental spills
  • Password protection for setup and calibration

Typical applications include: Ecological studies, environmental studies, chemical labs, food processing, titrations and quality assurance testing.

Delivery: Supplied with pH electrode (ECFG7370101B), ATC probe (PH5TEMB01P), RS232 cable, integral electrode holder, 100/240 VAC adapter and pH electrode refill solution.