FHA (from Bordetella pertussis, strain NCTC 10739)
Catalog # ENZOALX6301230100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:100 µG
- Source:Bordetella pertussis, strain NCTC 10739
- Seq:02
- Storage conditions:4 °C (long term)
- Protein synonyms:Filamentous hemagglutinin antigen
- Protein/peptide name:FHA
- Purity:>98%
- Colour:White
- Formulation:Lyophilised powder
- Tested applications:ELISA, Antibody capture, LINE assay
Specifications
About this item
Lyophilised from 0,05 M TRIS-HCl and 0,5 M sodium chloride at pH 8, 5% sucrose.