

(R)-(-)-2-Methylpiperazine ≥98%
Catalog # B22366.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:100.16 g/mol
Boiling Pt:155…156 °C
Melting Pt:76…78 °C
Flash Pt:65 °C (149 °F)
UN:2925
ADR:4.1,III
:GHS05
:GHS02
Storage Temperature:Ambient
CAS Number:75336-86-6
Formula:C₅H₁₂N₂
MDL Number:MFCD00192317