
(S)-(-)-1-Phenylethanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1445-91-6
:
Formula:C₈H₁₀O
MDL Number:MFCD00064264
:122.17 g/mol
Boiling Pt:207 °C (1013 hPa)
Density:1.018 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2937
ADR:6.1,III
:M-Clarity™ quality level = MQ100
:GHS07
Frequently Bought Together