

(S)-(-)-α-Methylbenzylamine
Catalog # MOLE14057640-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:121.18 g/mol
Merck Index:12,06106
Boiling Pt:187 °C (1013 hPa)
Melting Pt:–10 °C
Density:0.95 g/cm³ (20 °C)
EINECS:220-098-0
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:2627-86-3
Formula:C₈H₁₁N
MDL Number:MFCD00064406