

(S)-(-)-α-Methylbenzylamine ≥99%, ee 99.5%, ChiPros®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2627-86-3
:
Formula:C₈H₁₁N
MDL Number:MFCD00064406
:121.18 g/mol
Merck Index:14,6026
Boiling Pt:187 °C
Melting Pt:–10 °C
Density:0.948
Flash Pt:71 °C (159 °F)
Storage Temperature:Ambient
EINECS:220-098-0
UN:2922
ADR:8,II
:GHS06
:GHS05
Related Information
Frequently Bought Together