Order Entry
Austria
ContactUsLinkComponent
 
L(-)-Tyrosine
Catalog # A1677.1000
Supplier:  PanReac AppliChem
CAS Number:  
L(-)-Tyrosine
Catalog # A1677.1000
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :181.19 g/mol
Merck Index:13,09907
Boiling Pt:351 °C (1013 hPa)
Melting Pt:285 °C
Density:1.41 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-460-4
CAS Number:60-18-4
Formula:C₉H₁₁NO₃
MDL Number:MFCD00002606