L(+)-Histidine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:71-00-1
:
Formula:C₆H₉N₃O₂
MDL Number:MFCD00064315
:155.16 g/mol
Merck Index:13,04741
Melting Pt:272…273 °C
Density:1.44 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-745-3
Frequently Bought Together