(R)-(-)-3-Methyl-2-butanol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1572-93-6
:
Formula:C₅H₁₂O
MDL Number:MFCD00065949
:88.15 g/mol
Boiling Pt:112 °C
Density:0.810
Flash Pt:26 °C (78 °F)
Storage Temperature:Ambient
UN:1105
ADR:3,III
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together