L-Leucine, EMPROVE® EXPERT Ph. Eur., USP suitable for use as excipient, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
Synonyms: (S)-(+)-Leucine, Leu, H-Leu-OH, Leucine, L-Leucine
CAS Number:61-90-5
:
Formula:C₆H₁₃NO₂
MDL Number:MFCD00002617
:131.17 g/mol
Merck Index:14,05451
Melting Pt:300 °C
Density:1.165 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-522-0
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together