Order Entry
Austria
ContactUsLinkComponent
Parteck® SI 150 (Sorbitol) 97.0-100.5%, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, JSFA, E420, SAFC®
Parteck® SI 150 (Sorbitol) 97.0-100.5%, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, JSFA, E420, SAFC®
Catalog # 1.03583.9028
CAS Number:  
undefined
Parteck® SI 150 (Sorbitol) 97.0-100.5%, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, NF, JSFA, E420, SAFC®
Catalog # 1.03583.9028
CAS Number:  

Specifications

  • Pack type:
    Cardboard carton
  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • Assay (HPLC, calc.on anhydrous substance):
    97.0 - 100.5 %
  • Identity:
    passes test
  • pH-value (10 %; water):
    3.5 - 7.0
  • Free acid:
    passes test
  • Appearance of solution (25 %; water):
    passes test
  • Spec. rotation (α20/D, 10 %, borate complex, calc. on anhydrous substance):
    +4.0 - +7.0 °
  • Conductivity (25°C) (20 %; water):
    ≤ 20
  • Chloride (Cl):
    ≤ 0.002 %
  • Sulfate (SO₄):
    ≤ 0.006 %
  • Heavy metals (as Pb):
    ≤ 5 ppm
  • As (Arsenic):
    ≤ 1.3 ppm
  • Ni (Nickel) (#):
    ≤ 1 ppm
  • Pb (Lead):
    ≤ 0.5 ppm
  • Ru (Ruthenium) (#):
    ≤ 1 ppm
  • Related substances (HPLC, EP) (Major impurity (Mannitol)):
    ≤ 2.0 %
  • Related substances (HPLC, EP) (Sum of all impurities):
    ≤ 3.0 %
  • Reducing sugars after hydrolysis/total sugar (as glucose):
    ≤ 0.5
  • Reducing sugars (as glucose):
    ≤ 0.11 %
  • Sulfated ash:
    ≤ 0.02 %
  • Loss on drying:
    ≤ 2.0
  • Water:
    ≤ 1.5 %
  • Particle size (< 53 µm (270 mesh)):
    ≤ 5 %
  • Particle size (> 500 µm (35 mesh)):
    ≤ 5 %
  • Residual solvents (ICH Q3C):
    excluded by manufacturing process
  • Microbiological test:
    passes test
  • Colony count (aerobic bacteria (TAMC)):
    ≤ 5*10²
  • Colony count (Yeasts and moulds (TYMC)):
    ≤ 10²
  • Salmonella spp.:
    in 10g Substance not detectable
  • E.coli:
    in 1g substance not detectable
  • Staphylococcus aureus:
    in 1g substance not detectable
  • Pseudomonas aeruginosa:
    in 1g substance not detectable
  • Candida albicans:
    In 1g substance not detectable

Specification Test Results

About this item

 :
Merck Index:13,08797
Boiling Pt:295 °C (1013 hPa)
Melting Pt:94…98 °C
Density:1.524 g/cm³ (20 °C)
Flash Pt:>100 °C
 :182.17 g/mol
Storage Temperature:Ambient
EINECS:200-061-5
CAS Number:50-70-4
 :98201-93-5
Formula:HOH₂C(CH(OH))₄CH₂OH
MDL Number:MFCD00004708