Order Entry
Austria
ContactUsLinkComponent
Parteck® SI 450 (Sorbitol), EMPROVE® EXPERT NF, FCC, JSFA suitable for use as excipient, SAFC®
Parteck® SI 450 (Sorbitol), EMPROVE® EXPERT NF, FCC, JSFA suitable for use as excipient, SAFC®
Catalog # 1.03557.9028
CAS Number:  
undefined
Parteck® SI 450 (Sorbitol), EMPROVE® EXPERT NF, FCC, JSFA suitable for use as excipient, SAFC®
Catalog # 1.03557.9028
CAS Number:  

Specifications

  • Pack type:
    Cardboard carton
  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
Merck Index:13,08797
Boiling Pt:295 °C (1013 hPa)
Melting Pt:94…98 °C
Density:1.524 g/cm³ (20 °C)
Flash Pt:>100 °C
 :182.17 g/mol
Storage Temperature:Ambient
EINECS:200-061-5
CAS Number:50-70-4
 :98201-93-5
Formula:HOH₂C(CH(OH))₄CH₂OH
MDL Number:MFCD00004708