Order Entry
Austria
ContactUsLinkComponent
Parteck® SI 400 (Sorbitol), EMPROVE® EXPERT E420, Ph. Eur., NF, BP, JSFA suitable for use as excipient, SAFC®
Parteck® SI 400 (Sorbitol), EMPROVE® EXPERT E420, Ph. Eur., NF, BP, JSFA suitable for use as excipient, SAFC®
Catalog # 1.03140.9028
CAS Number:  
undefined
Parteck® SI 400 (Sorbitol), EMPROVE® EXPERT E420, Ph. Eur., NF, BP, JSFA suitable for use as excipient, SAFC®
Catalog # 1.03140.9028
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    25 kg
  • Seq:
    4283
  • Environmentally Preferable:

Specifications

About this item

Synonyms: (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
 :
Merck Index:13,08797
Boiling Pt:295 °C (1013 hPa)
Melting Pt:94…98 °C
Density:1.524 g/cm³ (20 °C)
Flash Pt:>100 °C
 :182.17 g/mol
Storage Temperature:Ambient
EINECS:200-061-5
CAS Number:50-70-4
 :98201-93-5
Formula:HOH₂C(CH(OH))₄CH₂OH
MDL Number:MFCD00004708