Order Entry
Austria
ContactUsLinkComponent
D-(-)-Sorbitol 97.0-100.5%, GPR RECTAPUR®
D-(-)-Sorbitol 97.0-100.5%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
D-(-)-Sorbitol 97.0-100.5%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (calculated on anhydrous):
    97.0- 100.5 %
  • Appearance of solution (10 %; water):
    Passes test
  • Identification:
    Passes test
  • Spec. opt. rot.(10 %; borate; anhydrous):
    4.0 - 7.0 °
  • Reducing sugars (calculated as C6H12O6):
    Max. 0.2 %
  • Ignition residue (SO4):
    Max. 0.1 %
  • Water:
    Max. 1.5 %
  • Cl (Chloride):
    Max. 50 ppm
  • SO4 (Sulphate):
    Max. 100 ppm
  • Ni (Nickel):
    Max. 1 ppm
  • Pb (Lead):
    Max. 0.5 ppm

Specification Test Results

Frequently Bought Together