Order Entry
Austria
Orders LinkContactUsLinkComponent
 

D-(-)-Mannitol, Millipore®

Supplier:   Merck
D-(-)-Mannitol, Millipore®
Supplier:  Merck
Certificates
D-(-)-Mannitol, Millipore®

3 Options Available Below

Product Details & Documents
Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064287
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,05769
  • Boiling Pt:290…295 °C (4 hPa)
  • Melting Pt:164…169 °C
  • Density:1.49 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Ambient
  • EINECS:200-711-8
Specifications for Product Family

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - 64,70 €
    64,70 €
    1 * 250 g
       64,70 €

    Specifications:
    More
    Specifications
    Cat. No.
    SIAL63560-250G-F
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 189,00 €
    189,00 €
    1 * 1 kg
       189,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SIAL63560-1KG-F
  • 5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - 658,00 €
    658,00 €
    1 * 5 kg
       658,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    SIAL63560-5KG-F