Order Entry
Austria
Orders LinkContactUsLinkComponent
D-(-)-Mannitol

D-(-)-Mannitol

5 kg
Catalog # MOLE71640365-5KG
Marketsource
Supplier:   Molekula
D-(-)-Mannitol
Catalog # MOLE71640365-5KG
Supplier:  Molekula
Marketsource
CAS Number:  69-65-8
D-(-)-Mannitol
D-(-)-Mannitol

5 kg

CAS Number:  69-65-8
Price
235,00 €
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064287
  • Molecular Weight:182.17 g/mol
  • Merck Index:13,05769
  • Boiling Pt:290…295 °C (4 hPa)
  • Melting Pt:164…169 °C
  • Density:1.49 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-711-8
Specifications

Specifications

  • Pk:
    5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.