


(S)-(+)-2-Aminooctane ≥99%, ee 99%, ChiPros®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:34566-04-6
:
Formula:C₈H₁₉N
MDL Number:MFCD01091021
Boiling Pt:163…165 °C
Melting Pt:< –20 °C
Density:0.770
Flash Pt:50 °C (122 °F)
UN:2734
ADR:8,II
:129.25 g/mol
:GHS06
:GHS05
:GHS02
Storage Temperature:Ambient
Related Information
Frequently Bought Together