L(+)-Potassium sodium tartrate tetrahydrate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
:6381-59-5
:
:KOOC(CHOH)₂COONa·4H₂O
:MFCD00150989
:282.22 g/mol
:13,07755
:70…80 °C
:∼1.767 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
:Ambient
:206-156-8
- Solubility:630 g/l (20 °C)
- Melting Point:70 - 80 °C
- Molar Mass:282.23 g/mol
- Bulk Density:1000 kg/m3
- Density:7.0 - 8.5 (50 g/l, H2O, 25 °C)
Frequently Bought Together