Order Entry
Austria
ContactUsLinkComponent
L(+)-Potassium sodium tartrate tetrahydrate 99+% for analysis
L(+)-Potassium sodium tartrate tetrahydrate 99+% for analysis
Certificates
undefined
L(+)-Potassium sodium tartrate tetrahydrate 99+% for analysis

Specification Test Results

  • Appearance:
    Semitransparent white Crystals
  • Infrared spectrum:
    Conforms
  • pH:
    5.5 to 8.5 (5 % at 20°C)
  • Titration with HClO4:
    99.0 to 102.0 %
  • Chloride (Cl):
    ≤50 ppm
  • Sulfate (SO4):
    ≤50 ppm
  • Calcium (Ca):
    ≤50 ppm
  • Iron (Fe):
    ≤10 ppm
  • Lead (Pb):
    ≤10 ppm

Specification Test Results

Related Information

Frequently Bought Together