Order Entry
Austria
ContactUsLinkComponent
L(+)-Tartaric acid ≥99.0% (by acidimetry) for resolution of racemates, for synthesis, Sigma-Aldrich®
Corrosive
L(+)-Tartaric acid ≥99.0% (by acidimetry) for resolution of racemates, for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
undefined
Corrosive
L(+)-Tartaric acid ≥99.0% (by acidimetry) for resolution of racemates, for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (acidimetric):
    ≥ 99.0 %
  • Spec. rotation [α²0/D (c=20 in water):
    +12.0 - +13.0 °
  • Identity (IR):
    passes test
  • Bulk Density:
    1000 kg/m3
  • Flash Point:
    210 °C
  • Density:
    1.76 g/cm3 (20 °C)
  • pH:
    1.6 (100 g/l, H2O, 25 °C)
  • Ignition Point:
    425 °C

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together