Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Decachlorobiphenyl (PCB No. 209) 10 µg/ml in isooctane, Dr Ehrenstorfer
Organic reference standard, Decachlorobiphenyl (PCB No. 209) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20020900IO
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, Decachlorobiphenyl (PCB No. 209) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20020900IO
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    2051-24-3
  • Description:
    Decachlorobiphenyl (PCB No. 209) 10 µg/ml in isooctane
  • Formula:
    C₁₂Cl₁₀
  • Description:
    Decachlorobiphenyl (PCB No. 209)

Specifications

About this item