Organic reference standard, 2,2',3,4,4',5,5',6-Octachlorobiphenyl (PCB No. 203) 100 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-203-CS
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:52663-76-0
- Description:2,2',3,4,4',5,5',6-Octachlorobiphenyl (PCB No. 203) 100 µg/ml in isooctane
- Formula:C₁₂H₂Cl₈
- Description:2,2',3,4,4',5,5',6-Octachlorobiphenyl (PCB No. 203)