Order Entry
Austria
ContactUsLinkComponent
L(+)-Tartaric acid, granules NF, FCC, J.T.Baker®
Corrosive
L(+)-Tartaric acid, granules NF, FCC, J.T.Baker®
Catalog # 4104-01
Supplier:  Avantor
CAS Number:  
undefined
Corrosive
L(+)-Tartaric acid, granules NF, FCC, J.T.Baker®
Catalog # 4104-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Poly bottle, white, blue T/E cap
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    Passes Test
  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • NF - Specific Rotation [Å]²⁵^D(calculated on dried basis)(+):
    12.0 - 13.0 Degree
  • NF - Loss on Drying:
    ≤0.5 %
  • NF - Residue on Ignition:
    ≤0.1 %
  • NF - Oxalate (C₂O₄):
    Passes Test
  • NF - Sulfate (SO₄):
    Passes Test
  • NF - Assay (HOCO(CHOH)₂COOH) (dried basis):
    99.7 - 100.5 %
  • FCC - Identification A:
    Passes Test
  • FCC - Identification B:
    Passes Test
  • FCC - Specific Rotation [Å]²⁵^D(calculated on dried basis)(+):
    12.0 - 13.0 Degree
  • FCC - Lead (Pb):
    ≤2 mg/kg
  • FCC - Loss on Drying:
    ≤0.5 %
  • FCC - Residue on Ignition:
    ≤0.1 %
  • FCC - Oxalate (C₂O₄):
    Passes Test
  • FCC - Sulfate (SO₄):
    Passes Test
  • FCC - Assay (HOCO(CHOH)₂COOH) (dried basis):
    99.7 - 100.5 %
  • Manufacturer Verification:
    Passes Test

Specification Test Results

About this item

 :
Merck Index:13,09158
Boiling Pt:399 °C (1013 hPa)
Melting Pt:168…170 °C
Density:1.759 g/cm³ (20 °C)
Flash Pt:150 °C (closed cup)
 :150.09 g/mol
Storage Temperature:Ambient
EINECS:201-766-0
REACH:01-2119537204-47
 :GHS05
CAS Number:87-69-4
Formula:C₄H₆O₆
MDL Number:MFCD00064207