Organic reference standard, 2,2',3,3',4,5,5',6,6'-Nonachlorobiphenyl (PCB No. 208) 100 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-208-CS
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:52663-77-1
- Description:2,2',3,3',4,5,5',6,6'-Nonachlorobiphenyl (PCB No. 208) 100 µg/ml in isooctane
- Formula:C₁₂H₂Cl₉
- Description:2,2',3,3',4,5,5',6,6'-Nonachlorobiphenyl (PCB No. 208)