Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, 2,4,4'-Trichlorobiphenyl (PCB No. 28) 10 µg/ml in isooctane, Dr Ehrenstorfer
Organic reference standard, 2,4,4'-Trichlorobiphenyl (PCB No. 28) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20002800IO
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 2,4,4'-Trichlorobiphenyl (PCB No. 28) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20002800IO
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    7012-37-5
  • Description:
    2,4,4'-Trichlorobiphenyl (PCB No. 28) 10 µg/ml in isooctane
  • Formula:
    C₁₂H₇Cl₃
  • Description:
    2,4,4'-Trichlorobiphenyl (PCB No. 28)

Specifications

About this item