Organic reference standard, 2,3',4,5-Tetrachlorobiphenyl (PCB No. 67) 35 µg/ml in isooctane, CIL
PROOCIL-PCB-67
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:73575-53-8
- Description:2,3',4,5-Tetrachlorobiphenyl (PCB No. 67) 35 µg/ml in isooctane
- Formula:C₁₂H₆Cl₄
- Description:2,3',4,5-Tetrachlorobiphenyl (PCB No. 67)