Organic reference standard, 2,3,3',4,6-Pentachlorobiphenyl (PCB No. 109) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-109
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:74472-35-8
- Description:2,3,3',4,6-Pentachlorobiphenyl (PCB No. 109) 35 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:2,3,3',4,6-Pentachlorobiphenyl (PCB No. 109)