Order Entry
Austria
ContactUsLinkComponent
L(+)-Aspartic acid, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
L(+)-Aspartic acid, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
Certificates
L(+)-Aspartic acid, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
Certificates

Specification Test Results

  • Assay (alkalimetric, calculated on dried substance):
    98.5 - 101.5 %
  • Identity (IR-spectrum):
    passes test
  • Identity (enantiomeric purity):
    passes test
  • Appearance:
    white to almost white, crystalline powder or colorless crystals
  • Appearance of solution (50 g/l; hydrochloric acid 1 mol/l):
    clear and not more intense in color than reference solution BY₆
  • Spec. rotation (α 20/D, 80 g/l, hydrochloric acid 6 mol/l, calc. on dried substance):
    +24.0 to +26.0
  • Chloride (Cl):
    ≤ 200 ppm
  • Sulfate (SO₄):
    ≤ 300 ppm
  • Fe (Iron):
    ≤ 10 ppm
  • Enantiomeric purity (HPLC) (D-aspartic acid):
    ≤ 0.3 %
  • Dicarboxylic acids (HPLC) (malic acid):
    ≤ 0.2 %
  • Dicarboxylic acids (HPLC) (fumaric acid):
    ≤ 0.10 %
  • Dicarboxylic acids (HPLC) (maleic acid):
    ≤ 0.10 %
  • Dicarboxylic acids (HPLC) (any unspecified impurity):
    ≤ 0.10 %
  • Dicarboxylic acids (HPLC) (total unspecified impurities):
    ≤ 0.3 %
  • Related compounds (HPLC) (maleic acid):
    ≤ 0.05 %
  • Related compounds (HPLC) (malic acid):
    ≤ 0.20 %
  • Related compounds (HPLC) (fumaric acid):
    ≤ 0.10 %
  • Related compounds (HPLC) (any unspecified impurity):
    ≤ 0.05 %
  • Related compounds (HPLC) (total unspecified impurities):
    ≤ 0.10 %
  • Ninhydrin-positive substances (LC)(impurity C (glutamic acid)):
    ≤ 0.2 %
  • Ninhydrin-positive substances (LC)(impurity D (alanine)):
    ≤ 0.2 %
  • Ninhydrin-positive substances (LC)(impurity G (asparagine)):
    ≤ 0.2 %
  • Ninhydrin-positive substances (LC) (any ninhydrin-positive impurity):
    ≤ 0.10 %
  • Ninhydrin-positive substances (LC) (total impurities):
    ≤ 1.0 %
  • Ninhydrin-positive substances (LC) (ammonium (570 nm)):
    ≤ 0.02 %
  • Residual solvents (ICH Q3C):
    excluded by manufacturing process
  • Sulfated ash (600 °C):
    ≤ 0.1 %
  • Loss on drying (105 °C; 3 h):
    ≤ 0.5 %

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together