Organic reference standard, 2,3,3',4,5,5'-Hexachlorobiphenyl (PCB No. 159) 100 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-159-CS
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:39635-35-3
- Description:2,3,3',4,5,5'-Hexachlorobiphenyl (PCB No. 159) 100 µg/ml in isooctane
- Formula:C₁₂H₄Cl₆
- Description:2,3,3',4,5,5'-Hexachlorobiphenyl (PCB No. 159)