Sodium gluconate USP, E576, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
CAS Number:527-07-1
:
Formula:HOH₂C(CH(OH))₄COONa
MDL Number:MFCD00064210
Merck Index:12,08766
Melting Pt:170…175 °C
:218.14 g/mol
Storage Temperature:Ambient
EINECS:208-407-7
Related Information
Certificates
Search by Lot NumberMERP8.17054.5000_TDS_EN ProductPage_817054
Product specification
msds_817054
SDS
Frequently Bought Together