Organic reference standard, 2,2',4,4',6-Pentachlorobiphenyl (PCB No. 100) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-100
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:39485-83-1
- Description:2,2',4,4',6-Pentachlorobiphenyl (PCB No. 100) 35 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:2,2',4,4',6-Pentachlorobiphenyl (PCB No