Organic reference standard, 2,2',3,4',5-Pentachlorobiphenyl (PCB No. 90) 10 µg/ml in isooctane, Dr Ehrenstorfer
EHERL20009000IO
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:68194-07-0
- Description:2,2',3,4',5-Pentachlorobiphenyl (PCB No. 90) 10 µg/ml in isooctane
- Formula:C₁₂H₅Cl₅
- Description:2,2',3,4',5-Pentachlorobiphenyl (PCB No. 90)