Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, 2,2',3,3',5-Pentachlorobiphenyl (PCB No. 83) 35 µg/ml in isooctane, CIL
Organic reference standard, 2,2',3,3',5-Pentachlorobiphenyl (PCB No. 83) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-83
CAS Number:  
Organic reference standard, 2,2',3,3',5-Pentachlorobiphenyl (PCB No. 83) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-83
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    60145-20-2
  • Description:
    2,2',3,3',5-Pentachlorobiphenyl (PCB No. 83) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₅Cl₅
  • Description:
    2,2',3,3',5-Pentachlorobiphenyl (PCB No. 83)

Specifications

About this item