Order Entry
Austria
ContactUsLinkComponent
L-Arginine hydrochloride
L-Arginine hydrochloride
Catalog # MOLE33349826-1KG
Supplier:  Molekula
CAS Number:  
L-Arginine hydrochloride
Catalog # MOLE33349826-1KG
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :210.66 g/mol
Merck Index:13,00785
Melting Pt:218 °C
Density:1.42 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:214-275-1
CAS Number:1119-34-2
Formula:C₆H₁₅ClN₄O₂
MDL Number:MFCD00064550