Order Entry
Austria
ContactUsLinkComponent
L-Arginine hydrochloride 98+%
L-Arginine hydrochloride 98+%
Certificates
undefined
L-Arginine hydrochloride 98+%
Certificates

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.5 % (105°C, 2 hrs)
  • Sulfated ash:
    ≤0.1 %
  • UV transmittance:
    ≥95 % (c=10, H2O, 430 nm) 1 cm cell
  • Specific optical rotation:
    +21.4° to +23.6° (20°C, 589 nm) (c=8, 6 N HCl)
  • Titration with HClO4:
    98.0 to 102.0 %
  • Sulfate (SO4):
    ≤300 ppm
  • Heavy metals (as Pb):
    ≤20 ppm
  • Other amino acids:
    ≤0.5 % (TLC)

Specification Test Results

Related Information

Frequently Bought Together