Organic reference standard, 2,3-Dichlorobiphenyl (PCB No. 5) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20000500IO
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:16605-91-7
- Description:2,3-Dichlorobiphenyl (PCB No. 5) 10 µg/ml in isooctane
- Formula:C₁₂H₈Cl₂
- Description:2,3-Dichlorobiphenyl (PCB No. 5