Order Entry
Austria
ContactUsLinkComponent
L(+)-Arginine 98.5-101.0%, EMPROVE® EXPERT Ph. Eur., JP, USP suitable for use as excipient, SAFC®
Irritant
L(+)-Arginine 98.5-101.0%, EMPROVE® EXPERT Ph. Eur., JP, USP suitable for use as excipient, SAFC®
Certificates
Irritant
L(+)-Arginine 98.5-101.0%, EMPROVE® EXPERT Ph. Eur., JP, USP suitable for use as excipient, SAFC®
Certificates

Specification Test Results

  • Assay (perchloric acid titration, calculated on dried substance):
    98.5 - 101.0 %
  • Identity (IR-spectrum):
    passes test
  • Appearance:
    white to almost white, crystalline powder or colorless crystals
  • Appearance of solution (50 g/l; water):
    clear and not more intense in color than reference solution BY₆
  • Appearance of solution (100 g/l, CO₂-free water):
    clear and colorless
  • Spec. rotation (α 20/D, 80 g/l, hydrochloric acid 250 g/l, calc. on dried substance):
    +25.5 to +28.5
  • Spec. rotation (α 25/D, 80 g/l, hydrochloric acid 6 mol/l, calc. on dried substance):
    +26.3 to +27.7
  • Spec. rotation (α 20/D, 80 g/l, hydrochloric acid 6 mol/l,previously dried substance):
    +26.9 to +27.9
  • pH (100 g/l, CO₂-free water):
    10.5 - 12.0
  • Chloride (Cl):
    ≤ 200 ppm
  • Sulfate (SO₄):
    ≤ 200 ppm
  • Heavy metals (as Pb):
    ≤ 10 ppm
  • Cu (Copper)*:
    ≤ 25 ppm
  • Fe (Iron):
    ≤ 10 ppm
  • Fe (Iron) (ICP):
    ≤ 10 ppm
  • Mn (Manganese):
    ≤ 25 ppm
  • Mo (Molybdenum)*:
    ≤ 2.5 ppm
  • Ni (Nickel)*:
    ≤ 2 ppm
  • Zn (Zinc):
    ≤ 130 ppm
  • Related Compounds, total (TLC):
    ≤ 0.5 %
  • Ninhydrin-positive substances (LC) (any ninhydrin-positive impurity):
    ≤ 0.2 %
  • Ninhydrin-positive substances (LC) (ammonium (570 nm)):
    ≤ 0.02 %
  • Ninhydrin-positive substances (LC) (total impurities):
    ≤ 0.5 %
  • Residual solvents (ICH Q3C):
    excluded by manufacturing process
  • Sulfated ash (600 °C):
    ≤ 0.1 %
  • Loss on drying (105 °C; 3 h):
    ≤ 0.30 %
  • Bacterial endotoxins:
    ≤ 2.0 I.U./g

Specification Test Results

Related Information

Frequently Bought Together