Organic reference standard, 2,2',3,3',5,5',6-Heptachlorobiphenyl (PCB No. 178) 100 µg/ml in isooctane, CIL
PROOCIL-PCB-178-CS
:
- Pack type:Glass bottle
- Pk:1,2 mL
- CAS No.:52663-67-9
- Description:2,2',3,3',5,5',6-Heptachlorobiphenyl (PCB No. 178) 100 µg/ml in isooctane
- Formula:C₁₂H₃Cl₇
- Description:2,2',3,3',5,5',6-Heptachlorobiphenyl (PCB No. 178)