Organic reference standard, 2,2',3,3',5,5',6,6'-Octachlorobiphenyl (PCB No. 202) 35 µg/ml in isooctane, CIL
Catalog # PROOCIL-PCB-202
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:2136-99-4
- Description:2,2',3,3',5,5',6,6'-Octachlorobiphenyl (PCB No. 202) 35 µg/ml in isooctane
- Formula:C₁₂H₂Cl₈
- Description:2,2',3,3',5,5',6,6'-Octachlorobiphenyl (PCB No. 202)