D-(-)-Diethyl tartrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: (2S,3S)-Diethyl 2,3-dihydroxysuccinate, (S,S)-(-)-Diethyl tartrate, (2S,3S)-(-)-Diethyl tartrate, (-)-Diethyl D-tartrate, (S,S)-(-)-Tartaric acid diethyl ester, D(-)-Tartaric acid diethyl ester
CAS Number:13811-71-7
:
Formula:C₈H₁₄O₆
MDL Number:MFCD00064451
Boiling Pt:280
Melting Pt:17
Density:1.2 g/cm³ (20 °C)
Molecular Weight:206.2 g/mol
:M-Clarity™ quality level = MQ100
EINECS:237-458-8
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together