Order Entry
Austria
ContactUsLinkComponent
 
Bio-17-ATP
Catalog # ENZOENZ42817
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Bio-17-ATP
Catalog # ENZOENZ42817
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    250 Nmo
  • Description:
    Biotin-17-adenosine-5’-triphosphate, 250 nmol
  • Seq:
    02

Specifications

About this item

Bio-17-ATP (Biotin-17-adenosine-5’-triphosphate) can replace ATP for in vitro transcription reaction catalysed by T3, T7 or SP6 RNA polymerases.

  • Appearance: Clear, colorless liquid
  • Purity: ≥93% (HPLC)
  • Concentration: 10 mM
  • Extinction coefficient: 18000 M⁻¹ cm⁻¹ (267 nm, pH 7,0)

The biotin-labelled RNA transcripts produced by these reactions are suitable for a wide range of applications such as nucleic acid hybridisation, sequencing, and genome analysis.

The biotin-labelled hybridised probes can be detected by a reporter molecule linked to streptavidin, avidin, or anit-biotin antibody. Such a complex can be detected directly, e.g. by excitation of a fluorophore conjugated to streptavidin, or indirectly, e.g. using an enzyme conjugate that can produce an insoluble coloured precipitate.