Nuclear receptor ligand library plates, SCREEN-WELL®
The SCREEN-WELL® Nuclear Receptor Ligand Library contains 74 compounds with defined, putative and potential activity at nuclear receptors.
- Stable for one year from the date of receipt when stored at –80 °C
- Shipped on dry ice
- Receptor agonists and antagonists are included
The library consists of a chemically diverse group of compounds which are supplied dissolved in DMSO at 10 mM (1 mM for 5 compounds) and aliquoted into deep-wel plates. The library is an ideal tool for chemical genomics, receptor de-orphaning and other routine pharmacological applications. The library includes ligands for the following classes of nuclear receptors: AHR, CAR, ER, FXR, LXR, PPAR, PXR, RAR, RXR, VDR, and other steroid receptors.
Frequently Bought Together