Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, 2,3,3',4,4',5,5'-Heptachlorobiphenyl (PCB No. 189) 35 µg/ml in isooctane, CIL
Organic reference standard, 2,3,3',4,4',5,5'-Heptachlorobiphenyl (PCB No. 189) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-189
 :  
Organic reference standard, 2,3,3',4,4',5,5'-Heptachlorobiphenyl (PCB No. 189) 35 µg/ml in isooctane, CIL
  PROOCIL-PCB-189
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    39635-31-9
  • Description:
    2,3,3',4,4',5,5'-Heptachlorobiphenyl (PCB No. 189) 35 µg/ml in isooctane
  • Formula:
    C₁₂H₃Cl₇
  • Description:
    2,3,3',4,4',5,5'-Heptachlorobiphenyl (PCB No. 189)